Learn More
3-Chloro-4-methoxypyridine, 98%, Thermo Scientific Chemicals
POA - £211.00
Chemical Identifiers
CAS | 89379-79-3 |
---|---|
Molecular Formula | C6H6ClNO |
Molecular Weight (g/mol) | 143.57 |
MDL Number | MFCD09743650 |
InChI Key | OFJIIJJCVDJQFW-UHFFFAOYSA-N |
Synonym | 4-methoxy-3-chloropyridine, 3-chloro-4-methoxy-pyridine, pyridine, 3-chloro-4-methoxy, pubchem6687 |
PubChem CID | 23436855 |
IUPAC Name | 3-chloro-4-methoxypyridine |
SMILES | COC1=C(Cl)C=NC=C1 |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
89379-79-3 | |
143.57 | |
OFJIIJJCVDJQFW-UHFFFAOYSA-N | |
23436855 | |
COC1=C(Cl)C=NC=C1 |
C6H6ClNO | |
MFCD09743650 | |
4-methoxy-3-chloropyridine, 3-chloro-4-methoxy-pyridine, pyridine, 3-chloro-4-methoxy, pubchem6687 | |
3-chloro-4-methoxypyridine |
Specifications
89379-79-3 | |
MFCD09743650 | |
4-methoxy-3-chloropyridine, 3-chloro-4-methoxy-pyridine, pyridine, 3-chloro-4-methoxy, pubchem6687 | |
COC1=C(Cl)C=NC=C1 | |
143.57 | |
143.57 | |
3-Chloro-4-methoxypyridine |
C6H6ClNO | |
116431 | |
OFJIIJJCVDJQFW-UHFFFAOYSA-N | |
3-chloro-4-methoxypyridine | |
23436855 | |
98% |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Keep cold
RUO – Research Use Only