missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-chloro-4-methyl-7-hydroxycoumarin, 97%, Thermo Scientific™
£29.81 - £115.81
Chemical Identifiers
CAS | 6174-86-3 |
---|---|
Molecular Formula | C10H7ClO3 |
Molecular Weight (g/mol) | 210.61 |
MDL Number | MFCD00006851 |
InChI Key | ODZHLDRQCZXQFQ-UHFFFAOYSA-N |
Synonym | chlorferone, 3-chloro-7-hydroxy-4-methyl-2h-chromen-2-one, 3-chloro-4-methyl-7-hydroxycoumarin, chlorferron, 3-chloro-7-hydroxy-4-methylcoumarin, 7-hydroxy-4-methyl-3-chlorocoumarin, chlorferon, 3-chloro-4-methylumbelliferone, 3-chloro-7-hydroxy-4-methyl-2-benzopyrone, 2h-1-benzopyran-2-one, 3-chloro-7-hydroxy-4-methyl |
PubChem CID | 5355079 |
ChEBI | CHEBI:38620 |
IUPAC Name | 3-chloro-7-hydroxy-4-methylchromen-2-one |
SMILES | CC1=C(Cl)C(=O)OC2=CC(O)=CC=C12 |
Description
Chemical Identifiers
6174-86-3 | |
210.61 | |
ODZHLDRQCZXQFQ-UHFFFAOYSA-N | |
5355079 | |
3-chloro-7-hydroxy-4-methylchromen-2-one |
C10H7ClO3 | |
MFCD00006851 | |
chlorferone, 3-chloro-7-hydroxy-4-methyl-2h-chromen-2-one, 3-chloro-4-methyl-7-hydroxycoumarin, chlorferron, 3-chloro-7-hydroxy-4-methylcoumarin, 7-hydroxy-4-methyl-3-chlorocoumarin, chlorferon, 3-chloro-4-methylumbelliferone, 3-chloro-7-hydroxy-4-methyl-2-benzopyrone, 2h-1-benzopyran-2-one, 3-chloro-7-hydroxy-4-methyl | |
CHEBI:38620 | |
CC1=C(Cl)C(=O)OC2=CC(O)=CC=C12 |
Specifications
6174-86-3 | |
100.0 | |
White | |
96% min. (HPLC) | |
MFCD00006851 | |
ODZHLDRQCZXQFQ-UHFFFAOYSA-N | |
3-chloro-7-hydroxy-4-methylchromen-2-one | |
5355079 | |
210.62 | |
Powder |
96.0 | |
246.0°C to 247.0°C | |
Authentic | |
C10H7ClO3 | |
chlorferone, 3-chloro-7-hydroxy-4-methyl-2h-chromen-2-one, 3-chloro-4-methyl-7-hydroxycoumarin, chlorferron, 3-chloro-7-hydroxy-4-methylcoumarin, 7-hydroxy-4-methyl-3-chlorocoumarin, chlorferon, 3-chloro-4-methylumbelliferone, 3-chloro-7-hydroxy-4-methyl-2-benzopyrone, 2h-1-benzopyran-2-one, 3-chloro-7-hydroxy-4-methyl | |
CC1=C(Cl)C(=O)OC2=CC(O)=CC=C12 | |
210.61 | |
CHEBI:38620 | |
97% | |
3-chloro-4-methyl-7-hydroxycoumarin |
Safety and Handling
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 228-217-8
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- 3-Chloro-4-methyl-7-hydroxycoumarin
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only