missing translation for 'onlineSavingsMsg'
Learn More
3-Chloro-5-(trifluoromethyl)pyridine-2-carboxylic acid, 97%, Thermo Scientific Chemicals Product Code.: 15448855

3-Chloro-5-(trifluoromethyl)pyridine-2-carboxylic acid, 97%, Thermo Scientific Chemicals

Product Code. 15448855
1 g
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 15448855

Brand: Thermo Scientific Alfa Aesar H34258.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80194-68-9
Molecular Formula C7H3ClF3NO2
Molecular Weight (g/mol) 225.551
MDL Number MFCD00277482
InChI Key HXRMCZBDTDCCOP-UHFFFAOYSA-N
Synonym 3-chloro-5-trifluoromethyl pyridine-2-carboxylic acid, 3-chloro-5-trifluoromethyl picolinic acid, 2-pyridinecarboxylic acid, 3-chloro-5-trifluoromethyl, 3-chloro-5-trifluoromethyl-pyridine-2-carboxylic acid, 3-chloro-5-trifluoromethyl-2-pyridinecarboxylic acid, 2-carboxy-3-chloro-5-trifluoromethyl pyridine, ksc446c7n, 3-chloro-5-trifluoromethylpicolinic acid, 3-chloro-5-trifluoromethyl-picolinic acidShow MoreShow Less
PubChem CID 2821908
IUPAC Name 3-chloro-5-(trifluoromethyl)pyridine-2-carboxylic acid
SMILES C1=C(C=NC(=C1Cl)C(=O)O)C(F)(F)F

Specifications

Melting Point 141°C to 143°C
Quantity 1 g
Formula Weight 225.6
Percent Purity 97%
Chemical Name or Material 3-Chloro-5-(trifluoromethyl)pyridine-2-carboxylic acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10178243 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberH34258.03
Lot Number81301184 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH34258.03
Lot Number10178243 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH34258.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Chloro-5-(trifluoromethyl)pyridine-2-carboxylic acid, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.