missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-chloroisonicotinic acid, 97%, Thermo Scientific™
£20.57 - £66.69
Chemical Identifiers
CAS | 88912-27-0 |
---|---|
Molecular Formula | C6H4ClNO2 |
Molecular Weight (g/mol) | 157.56 |
MDL Number | MFCD03094685 |
InChI Key | MYAZXWFEMDJTFE-UHFFFAOYSA-N |
Synonym | 3-chloroisonicotinic acid, 3-chloro-4-pyridinecarboxylic acid, 4-pyridinecarboxylic acid, 3-chloro, 3-chloro-4-pyridinecarboxylicacid, pubchem5087, acmc-209qxw, 3-chloroisonicotinic cid, 3-chloro-isonicotinic acid, ksc447s3l |
PubChem CID | 2735816 |
IUPAC Name | 3-chloropyridine-4-carboxylic acid |
SMILES | C1=CN=CC(=C1C(=O)O)Cl |
Description
Chemical Identifiers
88912-27-0 | |
157.56 | |
MYAZXWFEMDJTFE-UHFFFAOYSA-N | |
2735816 | |
C1=CN=CC(=C1C(=O)O)Cl |
C6H4ClNO2 | |
MFCD03094685 | |
3-chloroisonicotinic acid, 3-chloro-4-pyridinecarboxylic acid, 4-pyridinecarboxylic acid, 3-chloro, 3-chloro-4-pyridinecarboxylicacid, pubchem5087, acmc-209qxw, 3-chloroisonicotinic cid, 3-chloro-isonicotinic acid, ksc447s3l | |
3-chloropyridine-4-carboxylic acid |
Specifications
88912-27-0 | |
100.0 | |
Black to Brown | |
96% min. (HPLC) | |
MFCD03094685 | |
MYAZXWFEMDJTFE-UHFFFAOYSA-N | |
3-chloropyridine-4-carboxylic acid | |
2735816 | |
97% | |
3-Chloroisonicotinic acid |
96.0 | |
227°C to 229°C | |
Authentic | |
C6H4ClNO2 | |
3-chloroisonicotinic acid, 3-chloro-4-pyridinecarboxylic acid, 4-pyridinecarboxylic acid, 3-chloro, 3-chloro-4-pyridinecarboxylicacid, pubchem5087, acmc-209qxw, 3-chloroisonicotinic cid, 3-chloro-isonicotinic acid, ksc447s3l | |
C1=CN=CC(=C1C(=O)O)Cl | |
157.56 | |
157.56 | |
Solution |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present
GHS Signal Word: Warning
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- 3-Chloroisonicotinic acid
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only