missing translation for 'onlineSavingsMsg'
Learn More
3-Chlorophthalic anhydride, 95+%, Thermo Scientific Chemicals Product Code.: 10739023

3-Chlorophthalic anhydride, 95+%, Thermo Scientific Chemicals

Product Code. 10739023
5 g, Glass bottle
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10739023

Brand: Thermo Scientific Acros 381290050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 117-21-5
Molecular Formula C8H3ClO3
Molecular Weight (g/mol) 182.56
MDL Number MFCD00023107
InChI Key UERPUZBSSSAZJE-UHFFFAOYSA-N
Synonym 3-chlorophthalic anhydride, 4-chloroisobenzofuran-1,3-dione, 3-chlorophthalic acid anhydride, unii-vo7nxt0137, 1,3-isobenzofurandione, chloro, 4-chloro-1,3-dihydro-2-benzofuran-1,3-dione, 4-chloranyl-2-benzofuran-1,3-dione, 3clpa, pubchem19464, 3-chlorophthalicanhydrideShow MoreShow Less
PubChem CID 67014
IUPAC Name 4-chloro-2-benzofuran-1,3-dione
SMILES ClC1=CC=CC2=C1C(=O)OC2=O

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 123°C to 126°C
Color White or Yellow
Infrared Spectrum Authentic
Assay Percent Range 95+%
Packaging Glass bottle
Quantity 5 g
Formula Weight 182.56
Percent Purity 95+%
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material 3-Chlorophthalic anhydride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0411973 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number381290250,381290010,381290050
Lot NumberA0411973 Certificate TypeCertificate of Analysis Date08/06/2021 Catalog Number381290250,381290010,381290050
Lot NumberA0402786 Certificate TypeCertificate of Analysis Date28/08/2020 Catalog Number381290250,381290010,381290050

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Chlorophthalic anhydride, 95+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.