Learn More
3-Chlorotoluene, 97%, Thermo Scientific Chemicals
£33.20 - £278.17
Chemical Identifiers
CAS | 108-41-8 |
---|---|
Molecular Formula | C7H7Cl |
Molecular Weight (g/mol) | 126.58 |
MDL Number | MFCD00000595 |
InChI Key | OSOUNOBYRMOXQQ-UHFFFAOYSA-N |
Synonym | 3-chlorotoluene, m-chlorotoluene, benzene, 1-chloro-3-methyl, m-tolyl chloride, 3-chloro-1-methylbenzene, 1-methyl-3-chlorobenzene, toluene, m-chloro, meta-chlorotoluene, unii-5k8172k9tt, chlorotoluene 3- |
PubChem CID | 7931 |
IUPAC Name | 1-chloro-3-methylbenzene |
SMILES | CC1=CC=CC(Cl)=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10721241
|
Thermo Scientific Acros
160451000 |
100 mL | Glass bottle |
£33.20
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10468340
|
Thermo Scientific Acros
160452500 |
250 mL | Glass bottle |
£61.30
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10052010
|
Thermo Scientific Acros
160455000 |
500 mL | Glass bottle |
£129.00
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10656904
|
Thermo Scientific Acros
160450025 |
2.5 L | Glass bottle | N/A | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
108-41-8 | |
126.58 | |
OSOUNOBYRMOXQQ-UHFFFAOYSA-N | |
7931 | |
CC1=CC=CC(Cl)=C1 |
C7H7Cl | |
MFCD00000595 | |
3-chlorotoluene, m-chlorotoluene, benzene, 1-chloro-3-methyl, m-tolyl chloride, 3-chloro-1-methylbenzene, 1-methyl-3-chlorobenzene, toluene, m-chloro, meta-chlorotoluene, unii-5k8172k9tt, chlorotoluene 3- | |
1-chloro-3-methylbenzene |
Specifications
-48°C | |
96.0 | |
100.0 | |
160°C to 162°C | |
Liquid | |
97% | |
C7H7Cl | |
MFCD00000595 | |
15, 2175 | |
Solubility in water: slightly soluble | |
CC1=CC=CC(Cl)=C1 | |
126.58 | |
126.59 | |
3-Chlorotoluene, 97% |
108-41-8 | |
Colorless to Yellow | |
1.0700g/mL | |
Authentic | |
1.5210 to 1.523 | |
1.07 | |
CH3C6H4Cl | |
05, 291 | |
3-chlorotoluene, m-chlorotoluene, benzene, 1-chloro-3-methyl, m-tolyl chloride, 3-chloro-1-methylbenzene, 1-methyl-3-chlorobenzene, toluene, m-chloro, meta-chlorotoluene, unii-5k8172k9tt, chlorotoluene 3- | |
OSOUNOBYRMOXQQ-UHFFFAOYSA-N | |
1-chloro-3-methylbenzene | |
7931 | |
97% |
Safety and Handling
GHS H Statement
Toxic to aquatic life with long lasting effects.
Harmful if inhaled.
Flammable liquid and vapor.
GHS P Statement
Avoid release to the environment.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 203-580-5
missing translation for 'tsca' : TSCA
- 3-Chlorotoluene
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 2
- Flammable liquid Category 3
Hazard Statement
- H411-Toxic to aquatic life with long lasting effects.
- H332-Harmful if inhaled.
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P273-Avoid release to the environment.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
RUO – Research Use Only