Learn More
3-Cyclopentylpropionic acid, 98%, Thermo Scientific Chemicals
£15.40 - £100.00
Chemical Identifiers
CAS | 140-77-2 |
---|---|
Molecular Formula | C8H14O2 |
Molecular Weight (g/mol) | 142.20 |
MDL Number | MFCD00001392 |
InChI Key | ZRPLANDPDWYOMZ-UHFFFAOYSA-N |
Synonym | 3-cyclopentylpropionic acid, cyclopentanepropanoic acid, cyclopentanepropionic acid, 3-cylcopentylpropanoic acid, cyclopentylpropionic acid, propionic acid, 3-cyclopentyl, 3-cyclopentyl-propionic acid, cypionic acid, 3-cyclopentylpropionicacid, cipionate |
PubChem CID | 8818 |
ChEBI | CHEBI:50899 |
IUPAC Name | 3-cyclopentylpropanoic acid |
SMILES | OC(=O)CCC1CCCC1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10581652
|
Thermo Scientific Acros
111610050 |
5 g | Glass bottle |
£15.40
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10395930
|
Thermo Scientific
111610250 |
25 g | Glass bottle |
£39.10
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10437870
|
Thermo Scientific Acros
111611000 |
100 g | Glass bottle | ||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
140-77-2 | |
142.20 | |
ZRPLANDPDWYOMZ-UHFFFAOYSA-N | |
8818 | |
3-cyclopentylpropanoic acid |
C8H14O2 | |
MFCD00001392 | |
3-cyclopentylpropionic acid, cyclopentanepropanoic acid, cyclopentanepropionic acid, 3-cylcopentylpropanoic acid, cyclopentylpropionic acid, propionic acid, 3-cyclopentyl, 3-cyclopentyl-propionic acid, cypionic acid, 3-cyclopentylpropionicacid, cipionate | |
CHEBI:50899 | |
OC(=O)CCC1CCCC1 |
Specifications
140-77-2 | |
100.0 | |
0.9900g/mL | |
46°C | |
98% | |
C5H9CH2CH2CO2H | |
MFCD00001392 | |
3-cyclopentylpropionic acid, cyclopentanepropanoic acid, cyclopentanepropionic acid, 3-cylcopentylpropanoic acid, cyclopentylpropionic acid, propionic acid, 3-cyclopentyl, 3-cyclopentyl-propionic acid, cypionic acid, 3-cyclopentylpropionicacid, cipionate | |
ZRPLANDPDWYOMZ-UHFFFAOYSA-N | |
3-cyclopentylpropanoic acid | |
8818 | |
142.2 | |
Liquid |
97.5 | |
Colorless to Yellow | |
130.0°C to 132.0°C (12.0 mmHg) | |
Authentic | |
C8H14O2 | |
1.4560 to 1.4580 | |
0.99 | |
Solubility in water: insoluble | |
OC(=O)CCC1CCCC1 | |
142.20 | |
CHEBI:50899 | |
98% | |
3-Cyclopentylpropionic acid, 98% |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Flammable liquid and vapor.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 205-433-0
missing translation for 'tsca' : TSCA
RUO – Research Use Only