missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Ethoxy-2-cyclohexen-1-one, 99%, Thermo Scientific Chemicals
£34.70 - £185.00
Chemical Identifiers
CAS | 5323-87-5 |
---|---|
Molecular Formula | C8H12O2 |
Molecular Weight (g/mol) | 140.18 |
MDL Number | MFCD00001580 |
InChI Key | JWCFJPLIRVYENQ-UHFFFAOYSA-N |
Synonym | 3-ethoxy-2-cyclohexen-1-one, 3-ethoxycyclohex-2-enone, 2-cyclohexen-1-one, 3-ethoxy, 3-ethoxy-2-cyclohexenone, 3-ethoxy-2-cyclohexene-1-one, 3-ethoxycyclohex-2-ene-1-one, 1-ethoxycyclohexene-3-one, acmc-209l4y, 3-ethoxy-2-cyclohexen-one, 3-ethoxy-cyclohex-2-enone |
PubChem CID | 79216 |
IUPAC Name | 3-ethoxycyclohex-2-en-1-one |
SMILES | CCOC1=CC(=O)CCC1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10509430
|
Thermo Scientific Acros
117880100 |
10 g | Glass bottle |
£34.70
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10614962
|
Thermo Scientific Acros
117880250 |
25 g | Glass bottle |
£65.30
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10167563
|
Thermo Scientific Acros
117881000 |
100 g | Glass bottle |
£185.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
5323-87-5 | |
140.18 | |
JWCFJPLIRVYENQ-UHFFFAOYSA-N | |
79216 | |
CCOC1=CC(=O)CCC1 |
C8H12O2 | |
MFCD00001580 | |
3-ethoxy-2-cyclohexen-1-one, 3-ethoxycyclohex-2-enone, 2-cyclohexen-1-one, 3-ethoxy, 3-ethoxy-2-cyclohexenone, 3-ethoxy-2-cyclohexene-1-one, 3-ethoxycyclohex-2-ene-1-one, 1-ethoxycyclohexene-3-one, acmc-209l4y, 3-ethoxy-2-cyclohexen-one, 3-ethoxy-cyclohex-2-enone | |
3-ethoxycyclohex-2-en-1-one |
Specifications
5323-87-5 | |
100.0 | |
1.0400g/mL | |
107°C | |
98.5% min. (GC) | |
C2H5OC6H7(=O) | |
MFCD00001580 | |
3-ethoxy-2-cyclohexen-1-one, 3-ethoxycyclohex-2-enone, 2-cyclohexen-1-one, 3-ethoxy, 3-ethoxy-2-cyclohexenone, 3-ethoxy-2-cyclohexene-1-one, 3-ethoxycyclohex-2-ene-1-one, 1-ethoxycyclohexene-3-one, acmc-209l4y, 3-ethoxy-2-cyclohexen-one, 3-ethoxy-cyclohex-2-enone | |
JWCFJPLIRVYENQ-UHFFFAOYSA-N | |
3-ethoxycyclohex-2-en-1-one | |
79216 | |
99% | |
3-Ethoxy-2-cyclohexen-1-one |
98.5 | |
Peach to White | |
76°C to 78°C (1.0 mmHg) | |
Authentic | |
C8H12O2 | |
1.5030 to 1.505 | |
1.04 | |
Solubility in water: slightly soluble | |
CCOC1=CC(=O)CCC1 | |
140.18 | |
140.18 | |
Crystalline Powder |
Safety and Handling
missing translation for 'einecsNumber' : 226-190-7
RUO – Research Use Only