missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methyl-1-butanol, 99%, for biochemistry, AcroSeal™, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mL
1 L
2.5 L
Packaging:
AcroSeal™ Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | -70.0°C |
Density | 0.8000g/mL |
Boiling Point | 130.0°C |
Flash Point | 45°C |
Assay Percent Range | 98.5% min. (GC) |
Packaging | AcroSeal™ Glass bottle |
Linear Formula | (CH3)2CHCH2CH2OH |
Refractive Index | 1.4050 to 1.4070 |
Quantity | 1 L |
Beilstein | 01,392 |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0466756 | Certificate of Analysis | 17/10/2024 | 412720010,412721000,412720025 |
A0458487 | Certificate of Analysis | 20/12/2023 | 412720010,412721000,412720025 |
A0452906 | Certificate of Analysis | 16/05/2023 | 412720010,412721000,412720025 |
A0450970 | Certificate of Analysis | 02/05/2023 | 412720010,412721000,412720025 |
A0431963 | Certificate of Analysis | 05/01/2023 | 412720010,412721000,412720025 |
Safety and Handling


Product Identifier
- 3-Methyl-1-butanol
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H332-Harmful if inhaled.
- H226-Flammable liquid and vapour.
- EUH066-Repeated exposure may cause skin dryness or cracking.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
3-Methyl-1-butanol, 99%, for biochemistry, AcroSeal™, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction