missing translation for 'onlineSavingsMsg'
Learn More
3-Methyl-1-butanol, ACS reagent, Thermo Scientific Chemicals Product Code.: 10330244

3-Methyl-1-butanol, ACS reagent, Thermo Scientific Chemicals

Product Code. 10330244
1 L, Glass bottle
Click to view available options
Quantity:
500 mL
1 L
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10330244

Brand: Thermo Scientific Acros 412730010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-51-3
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
MDL Number MFCD00002934
InChI Key PHTQWCKDNZKARW-UHFFFAOYSA-N
Synonym 3-methyl-1-butanol, isoamyl alcohol, isopentyl alcohol, isopentanol, 3-methylbutanol, 1-butanol, 3-methyl, isoamylol, isobutylcarbinol, iso-amylalkohol, isobutyl carbinolShow MoreShow Less
PubChem CID 31260
ChEBI CHEBI:15837
IUPAC Name 3-methylbutan-1-ol
SMILES CC(C)CCO

Specifications

Melting Point -70.0°C
Density 0.8000g/mL
Boiling Point 130.0°C
Flash Point 45°C
Packaging Glass bottle
Linear Formula (CH3)2CHCH2CH2OH
Refractive Index 1.4050 to 1.4070
Quantity 1 L
Beilstein 01, 392
Residue after Evaporation 0.003% max.
Merck Index 15, 5241
Specific Gravity 0.8
Solubility Information Solubility in water: 25g/L (20°C). Other solubilities: miscible with alcohol,ether,benzene,oils,,chloroform
Viscosity 4.3 mPa.s (20°C)
Formula Weight 88.15
Percent Purity ≥98.5%
Grade ACS Reagent
Titratable Acid 0.002 meq/g max
Physical Form Liquid
Chemical Name or Material 3-Methyl-1-butanol, 98.5%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0467664 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number412730025,412735000,412730010
Lot NumberA0467663 Certificate TypeCertificate of Analysis Date30/01/2025 Catalog Number412730025,412735000,412730010
Lot NumberA0465533 Certificate TypeCertificate of Analysis Date28/08/2024 Catalog Number412730025,412735000,412730010
Lot NumberA0460840 Certificate TypeCertificate of Analysis Date17/04/2024 Catalog Number412730025,412735000,412730010
Lot NumberA0455004 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number412730025,412735000,412730010
compliance-icons compliance-icons
Product Identifier
  • 3-Methyl-1-butanol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Methyl-1-butanol, ACS reagent, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.