Learn More
3-Methyl-2-pyridone, 97+%, Thermo Scientific™
£15.00 - £125.00
Chemical Identifiers
CAS | 1003-56-1 |
---|---|
Molecular Formula | C6H7NO |
Molecular Weight (g/mol) | 109.128 |
MDL Number | MFCD00039704 |
InChI Key | MVKDNXIKAWKCCS-UHFFFAOYSA-N |
Synonym | 3-methylpyridin-2-ol, 2-hydroxy-3-methylpyridine, 3-methyl-2-pyridone, 3-methylpyridin-2 1h-one, 3-methyl-2-pyridinol, 2 1h-pyridinone, 3-methyl, 3-methyl-2-pyridinone, unii-vm19fn4hhv, 3-methyl-2-hydroxypyridine, 3-methyl-2 1h-pyridinone |
PubChem CID | 70481 |
IUPAC Name | 3-methyl-1H-pyridin-2-one |
SMILES | CC1=CC=CNC1=O |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications3-Methyl-2-pyridone, is used as a reactant with chloro-methoxy-methane to produce 1-methoxymethyl-3-methyl-1H-pyridin-2-one. This reaction will need reagent solvent CH2Cl2. The reaction time is 24 hours with ambient temperature
Solubility
Soluble in methanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
Chemical Identifiers
1003-56-1 | |
109.128 | |
MVKDNXIKAWKCCS-UHFFFAOYSA-N | |
70481 | |
CC1=CC=CNC1=O |
C6H7NO | |
MFCD00039704 | |
3-methylpyridin-2-ol, 2-hydroxy-3-methylpyridine, 3-methyl-2-pyridone, 3-methylpyridin-2 1h-one, 3-methyl-2-pyridinol, 2 1h-pyridinone, 3-methyl, 3-methyl-2-pyridinone, unii-vm19fn4hhv, 3-methyl-2-hydroxypyridine, 3-methyl-2 1h-pyridinone | |
3-methyl-1H-pyridin-2-one |
Specifications
1003-56-1 | |
C6H7NO | |
1422044 | |
Soluble in methanol. | |
CC1=CC=CNC1=O | |
109.128 | |
109.13 | |
3-Methyl-2-pyridone |
137°C to 142°C | |
MFCD00039704 | |
3-methylpyridin-2-ol, 2-hydroxy-3-methylpyridine, 3-methyl-2-pyridone, 3-methylpyridin-2 1h-one, 3-methyl-2-pyridinol, 2 1h-pyridinone, 3-methyl, 3-methyl-2-pyridinone, unii-vm19fn4hhv, 3-methyl-2-hydroxypyridine, 3-methyl-2 1h-pyridinone | |
MVKDNXIKAWKCCS-UHFFFAOYSA-N | |
3-methyl-1H-pyridin-2-one | |
70481 | |
≥97% |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 213-710-2
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures