Learn More
3-Methyl-3-oxetanemethanol, 97%, Thermo Scientific Chemicals
Description
3-Methyl-3-oxetanemethanol has been used in the preparation of star-shaped copolymer consisting of a hyperbranched poly(3-methyl-3-oxetanemethanol) core and polytetrahydrofuran arms, pyridyl disulfide-functionalized cyclic carbonate monomer, required for the synthesis of of functional poly(ε-caprolactone) containing pendant pyridyl disulphide groups and δ-lactams. It is used to synthesize octahydroindole alkaloids and γ-butenolides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
3-Methyl-3-oxetanemethanol has been used in the preparation of star-shaped copolymer consisting of a hyperbranched poly(3-methyl-3-oxetanemethanol) core and polytetrahydrofuran arms, pyridyl disulfide-functionalized cyclic carbonate monomer, required for the synthesis of of functional poly(ε-caprolactone) containing pendant pyridyl disulphide groups and δ-lactams. It is used to synthesize octahydroindole alkaloids and γ-butenolides.
Solubility
Soluble in chloroform, methanol.
Notes
Protect from heat. Store away from oxidizing agents. Store at 4°C.

Specifications
Specifications
Density | 1.024 |
Boiling Point | 79°C to 80°C (40 mmHg) |
Flash Point | 98°C (208°F) |
Refractive Index | 1.446 |
Quantity | 25 g |
Beilstein | 102773 |
Solubility Information | Soluble in chloroform,methanol. |
Formula Weight | 102.13 |
Percent Purity | 97% |
Chemical Name or Material | 3-Methyl-3-oxetanemethanol |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
10172505 | Certificate of Analysis | 23/11/2021 | B25111.22,B25111.14 |
10227306 | Certificate of Analysis | 23/11/2021 | B25111.22,B25111.14 |
10229403 | Certificate of Analysis | 23/11/2021 | B25111.22,B25111.14 |
10178268 | Certificate of Analysis | 23/11/2021 | B25111.22,B25111.14 |
10151655 | Certificate of Analysis | 23/11/2021 | B25111.22,B25111.14 |
Safety and Handling

- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.