missing translation for 'onlineSavingsMsg'
Learn More
3-Methyl-3-oxetanemethanol, 97%, Thermo Scientific Chemicals Product Code.: 11339889

3-Methyl-3-oxetanemethanol, 97%, Thermo Scientific Chemicals

Product Code. 11339889
100 g
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy

Product Code. 11339889

Brand: Thermo Scientific Alfa Aesar B25111.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

3-Methyl-3-oxetanemethanol has been used in the preparation of star-shaped copolymer consisting of a hyperbranched poly(3-methyl-3-oxetanemethanol) core and polytetrahydrofuran arms, pyridyl disulfide-functionalized cyclic carbonate monomer, required for the synthesis of of functional poly(ε-caprolactone) containing pendant pyridyl disulphide groups and δ-lactams. It is used to synthesize octahydroindole alkaloids and γ-butenolides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methyl-3-oxetanemethanol has been used in the preparation of star-shaped copolymer consisting of a hyperbranched poly(3-methyl-3-oxetanemethanol) core and polytetrahydrofuran arms, pyridyl disulfide-functionalized cyclic carbonate monomer, required for the synthesis of of functional poly(ε-caprolactone) containing pendant pyridyl disulphide groups and δ-lactams. It is used to synthesize octahydroindole alkaloids and γ-butenolides.

Solubility
Soluble in chloroform, methanol.

Notes
Protect from heat. Store away from oxidizing agents. Store at 4°C.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3143-02-0
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.133
MDL Number MFCD00010273
InChI Key NLQMSBJFLQPLIJ-UHFFFAOYSA-N
Synonym 3-methyl-3-oxetanemethanol, 3-methyloxetan-3-yl methanol, 3-hydroxymethyl-3-methyloxetane, 3-oxetanemethanol, 3-methyl, 3-methyl-3-oxetanyl methanol, 3-methyloxetane-3-methanol, 3-methyl-3-oxethanemethanol, 3-methyl-3-hydroxymethyloxetane, 3-hydroxymethyl-3-methyl-oxetane, 3-methyl-oxetan-3-yl-methanolShow MoreShow Less
PubChem CID 137837
IUPAC Name (3-methyloxetan-3-yl)methanol
SMILES CC1(COC1)CO

Specifications

Density 1.024
Boiling Point 79°C to 80°C (40 mmHg)
Flash Point 98°C (208°F)
Refractive Index 1.446
Quantity 100 g
Beilstein 102773
Solubility Information Soluble in chloroform,methanol.
Formula Weight 102.13
Percent Purity 97%
Chemical Name or Material 3-Methyl-3-oxetanemethanol

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10172505 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB25111.22,B25111.14
Lot Number10227306 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB25111.22,B25111.14
Lot Number10178268 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB25111.22,B25111.14
Lot Number10229403 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB25111.22,B25111.14
Lot Number10151655 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB25111.22,B25111.14
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Methyl-3-oxetanemethanol, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.