Learn More
3-Methylcyclohexanone, 97%, Thermo Scientific Chemicals
£19.00 - £120.00
Identificadores de productos químicos
CAS | 591-24-2 |
---|---|
Molecular Formula | C7H12O |
Molecular Weight (g/mol) | 112.17 |
MDL Number | MFCD00001639 |
InChI Key | UJBOOUHRTQVGRU-UHFFFAOYNA-N |
Synonym | 3-methylcyclohexanone, cyclohexanone, 3-methyl, methyl-3 cyclohexanone-1, 3-methyl cyclohexanone, methyl-3 cyclohexanone-1 french, m-methylcyclohexanone, r-3-methylcyclohexanone, +-3r-3-methylcyclohexanone, cyclohexanone, 3-methyl-, r, cyclohexanone, 3-methyl-, s |
PubChem CID | 11567 |
IUPAC Name | 3-methylcyclohexan-1-one |
SMILES | CC1CCCC(=O)C1 |
Código de producto | Marca | Quantity | Packaging | Precio | Cantidad y disponibilidad | ||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Quantity | Packaging | Precio | Cantidad y disponibilidad | ||||
10366480
|
Thermo Scientific Acros
126630050 |
5 g | Glass bottle |
£19.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10543291
|
Thermo Scientific Acros
126631000 |
100 g | Glass bottle |
£36.20
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10459050
|
Thermo Scientific Acros
126635000 |
500 g | Glass bottle |
£120.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Descripción
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Identificadores de productos químicos
591-24-2 | |
112.17 | |
UJBOOUHRTQVGRU-UHFFFAOYNA-N | |
11567 | |
CC1CCCC(=O)C1 |
C7H12O | |
MFCD00001639 | |
3-methylcyclohexanone, cyclohexanone, 3-methyl, methyl-3 cyclohexanone-1, 3-methyl cyclohexanone, methyl-3 cyclohexanone-1 french, m-methylcyclohexanone, r-3-methylcyclohexanone, +-3r-3-methylcyclohexanone, cyclohexanone, 3-methyl-, r, cyclohexanone, 3-methyl-, s | |
3-methylcyclohexan-1-one |
Especificaciones
591-24-2 | |
100.0 | |
Colorless to Yellow | |
169.0°C to 170.0°C | |
Authentic | |
C7H12O | |
1.4440 to 1.4460 | |
07, 15 | |
3-methylcyclohexanone, cyclohexanone, 3-methyl, methyl-3 cyclohexanone-1, 3-methyl cyclohexanone, methyl-3 cyclohexanone-1 french, m-methylcyclohexanone, r-3-methylcyclohexanone, +-3r-3-methylcyclohexanone, cyclohexanone, 3-methyl-, r, cyclohexanone, 3-methyl-, s | |
UJBOOUHRTQVGRU-UHFFFAOYNA-N | |
3-methylcyclohexan-1-one | |
11567 | |
97% | |
3-Methylcyclohexanone, 97% |
96.0 | |
-73.0°C | |
0.9150g/mL | |
48°C | |
97% | |
CH3C6H9(=O) | |
MFCD00001639 | |
0.915 | |
Solubility in water: insoluble | |
CC1CCCC(=O)C1 | |
112.17 | |
112.17 | |
Liquid |
Seguridad y manipulación
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapor.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF INHALED: Remove to f
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 209-710-7
missing translation for 'rtecsNumber' : GW1750100
missing translation for 'tsca' : TSCA
- 3-Methylcyclohexanone
Palabra de advertencia
- Warning
Clasificación
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Indicaciones de peligro
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H226-Flammable liquid and vapour.
Consejos de prudencia
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.