missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(3-Methylphenyl)methanethiol, 95%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
25 g
Specifications
Specifications
CAS Min % | 95.0 |
Assay Percent Range | 95% |
Packaging | Amber glass bottle |
Linear Formula | CH3C6H4CH2SH |
Quantity | 10 g |
Synonym | 3-methylphenyl methanethiol, 3-methylbenzyl mercaptan, m-methylbenzyl mercaptan, 3-methylbenzenemethanethiol, m-methylbenzylmercaptan, acmc-1cq7k, 3-methyl-benzenemethanethiol, benzenemethanethiol,3-methyl, benzenemethanethiol, 3-methyl, 3-methylphenyl methane-1-thiol |
Formula Weight | 138.23 |
Percent Purity | 95% |
Chemical Name or Material | (3-Methylphenyl)methanethiol |
Product Suggestions
Customers who viewed this item also viewed.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
(3-Methylphenyl)methanethiol, 95%, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction