Learn More
3-(Methylthio)phenyl isocyanate, 96%, Thermo Scientific Chemicals
£35.20 - £125.00
Chemical Identifiers
CAS | 28479-19-8 |
---|---|
Molecular Formula | C8H7NOS |
Molecular Weight (g/mol) | 165.21 |
MDL Number | MFCD00013863 |
InChI Key | BKJABLMNBSVKCV-UHFFFAOYSA-N |
Synonym | 3-methylthio phenyl isocyanate, 1-isocyanato-3-methylthio benzene, 3-methylthiophenyl isocyanate, 1-isocyanato-3-methylsulfanyl benzene, 1-isocyanato-3-methylsulfanyl-benzene, 3-methylthio phenylisocyanate, benzene, 1-isocyanato-3-methylthio, 3-methylthiobenzenisocyanate, acmc-20anig, pubchem14329 |
PubChem CID | 141418 |
IUPAC Name | 1-isocyanato-3-methylsulfanylbenzene |
SMILES | CSC1=CC=CC(=C1)N=C=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11368555
|
Thermo Scientific Alfa Aesar
L12008.03 |
1 g |
£35.20
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11378555
|
Thermo Scientific Alfa Aesar
L12008.06 |
5 g |
£125.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
3-(Methylthio)phenyl isocyanate is used as a coupling reagent, N-protecting reagent and as gas chromatography derivatizing reagent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-(Methylthio)phenyl isocyanate is used as a coupling reagent, N-protecting reagent and as gas chromatography derivatizing reagent.
Solubility
Hydrolyzes in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents, amines and alcohols. It is sensitive to moisture. Store under dry inert gas.
Chemical Identifiers
28479-19-8 | |
165.21 | |
BKJABLMNBSVKCV-UHFFFAOYSA-N | |
141418 | |
CSC1=CC=CC(=C1)N=C=O |
C8H7NOS | |
MFCD00013863 | |
3-methylthio phenyl isocyanate, 1-isocyanato-3-methylthio benzene, 3-methylthiophenyl isocyanate, 1-isocyanato-3-methylsulfanyl benzene, 1-isocyanato-3-methylsulfanyl-benzene, 3-methylthio phenylisocyanate, benzene, 1-isocyanato-3-methylthio, 3-methylthiobenzenisocyanate, acmc-20anig, pubchem14329 | |
1-isocyanato-3-methylsulfanylbenzene |
Specifications
28479-19-8 | |
129°C to 130°C (5 mmHg) | |
C8H7NOS | |
MFCD00013863 | |
387507 | |
3-methylthio phenyl isocyanate, 1-isocyanato-3-methylthio benzene, 3-methylthiophenyl isocyanate, 1-isocyanato-3-methylsulfanyl benzene, 1-isocyanato-3-methylsulfanyl-benzene, 3-methylthio phenylisocyanate, benzene, 1-isocyanato-3-methylthio, 3-methylthiobenzenisocyanate, acmc-20anig, pubchem14329 | |
BKJABLMNBSVKCV-UHFFFAOYSA-N | |
1-isocyanato-3-methylsulfanylbenzene | |
141418 | |
96% |
1.199 | |
113°C (235°F) | |
1.603 | |
UN2206 | |
Moisture sensitive | |
Hydrolyzes in water. | |
CSC1=CC=CC(=C1)N=C=O | |
165.21 | |
165.22 | |
3-(Methylthio)phenyl isocyanate |
Safety and Handling
GHS H Statement
H331-H334-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P272-P280g-P301+P312-P302+P352-P304+P340-P312-P330-P333+P313-P363-P501c
H302-H317-H335
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ISOCYANATES, TOXIC, N.O.S.
missing translation for 'einecsNumber' : 000-000-0
missing translation for 'tsca' : No
Recommended Storage : Keep cold
RUO – Research Use Only