missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-morpholin-4-ylaniline, 97%, Thermo Scientific™
£12.75 - £133.00
Chemical Identifiers
CAS | 159724-40-0 |
---|---|
Molecular Formula | C10H14N2O |
Molecular Weight (g/mol) | 178.24 |
MDL Number | MFCD03197165 |
InChI Key | ZJWLMZURLIHVHE-UHFFFAOYSA-N |
Synonym | 3-4-morpholinyl aniline, 3-morpholinoaniline, 3-morpholinobenzenamine, 3-morpholin-4-yl aniline, benzenamine, 3-4-morpholinyl, 3-morpholin-4-yl-phenylamine, 4-3-aminophenyl morpholine, 3-morpholin-4-ylphenylamine, 3-morpholinylaniline, meta-morpholino aniline |
PubChem CID | 847768 |
IUPAC Name | 3-morpholin-4-ylaniline |
SMILES | NC1=CC=CC(=C1)N1CCOCC1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10779944
|
Thermo Scientific Maybridge
CC33814CB |
250 mg | Amber Glass Bottle |
£12.75
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10084612
|
Thermo Scientific Maybridge
CC33814DA |
1 g | Amber Glass Bottle |
£33.40
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10173714
|
Thermo Scientific Maybridge
CC33814DE |
5 g | Amber Glass Bottle |
£133.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
159724-40-0 | |
178.24 | |
ZJWLMZURLIHVHE-UHFFFAOYSA-N | |
847768 | |
NC1=CC=CC(=C1)N1CCOCC1 |
C10H14N2O | |
MFCD03197165 | |
3-4-morpholinyl aniline, 3-morpholinoaniline, 3-morpholinobenzenamine, 3-morpholin-4-yl aniline, benzenamine, 3-4-morpholinyl, 3-morpholin-4-yl-phenylamine, 4-3-aminophenyl morpholine, 3-morpholin-4-ylphenylamine, 3-morpholinylaniline, meta-morpholino aniline | |
3-morpholin-4-ylaniline |
Specifications
159724-40-0 | |
C10H14N2O | |
3-4-morpholinyl aniline, 3-morpholinoaniline, 3-morpholinobenzenamine, 3-morpholin-4-yl aniline, benzenamine, 3-4-morpholinyl, 3-morpholin-4-yl-phenylamine, 4-3-aminophenyl morpholine, 3-morpholin-4-ylphenylamine, 3-morpholinylaniline, meta-morpholino aniline | |
NC1=CC=CC(=C1)N1CCOCC1 | |
178.24 | |
178.23 | |
3-Morpholin-4-ylaniline |
97% | |
MFCD03197165 | |
ZJWLMZURLIHVHE-UHFFFAOYSA-N | |
3-morpholin-4-ylaniline | |
847768 | |
97% |