Learn More
3-Nitrobenzyl chloride, 99%, Thermo Scientific™
£73.23 - £232.65
Chemical Identifiers
CAS | 619-23-8 |
---|---|
Molecular Formula | C7H6ClNO2 |
Molecular Weight (g/mol) | 171.58 |
MDL Number | MFCD00007272 |
InChI Key | APGGSERFJKEWFG-UHFFFAOYSA-N |
Synonym | 3-nitrobenzyl chloride, 1-chloromethyl-3-nitrobenzene, m-nitrobenzyl chloride, alpha-chloro-3-nitrotoluene, benzene, 1-chloromethyl-3-nitro, toluene, alpha-chloro-m-nitro, .alpha.-chloro-m-nitrotoluene, unii-ojl133898o, 3-nitrobenzylchloride, ccris 2324 |
PubChem CID | 12078 |
IUPAC Name | 1-(chloromethyl)-3-nitrobenzene |
SMILES | [O-][N+](=O)C1=CC=CC(CCl)=C1 |
Description
Chemical Identifiers
619-23-8 | |
171.58 | |
APGGSERFJKEWFG-UHFFFAOYSA-N | |
12078 | |
[O-][N+](=O)C1=CC=CC(CCl)=C1 |
C7H6ClNO2 | |
MFCD00007272 | |
3-nitrobenzyl chloride, 1-chloromethyl-3-nitrobenzene, m-nitrobenzyl chloride, alpha-chloro-3-nitrotoluene, benzene, 1-chloromethyl-3-nitro, toluene, alpha-chloro-m-nitro, .alpha.-chloro-m-nitrotoluene, unii-ojl133898o, 3-nitrobenzylchloride, ccris 2324 | |
1-(chloromethyl)-3-nitrobenzene |
Specifications
619-23-8 | |
100.0 | |
Brown to Yellow | |
>110°C | |
99% | |
O2NC6H4CH2Cl | |
3-nitrobenzyl chloride, 1-chloromethyl-3-nitrobenzene, m-nitrobenzyl chloride, alpha-chloro-3-nitrotoluene, benzene, 1-chloromethyl-3-nitro, toluene, alpha-chloro-m-nitro, .alpha.-chloro-m-nitrotoluene, unii-ojl133898o, 3-nitrobenzylchloride, ccris 2324 | |
[O-][N+](=O)C1=CC=CC(CCl)=C1 | |
171.58 | |
171.58 | |
Crystalline Solid |
98.5 | |
41°C to 46°C | |
85°C to 87°C (5.0 mmHg) | |
Authentic | |
C7H6ClNO2 | |
MFCD00007272 | |
APGGSERFJKEWFG-UHFFFAOYSA-N | |
1-(chloromethyl)-3-nitrobenzene | |
12078 | |
99% | |
3-Nitrobenzyl chloride, 99% |
Safety and Handling
GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 210-586-1
missing translation for 'rtecsNumber' : XS9090000
missing translation for 'tsca' : TSCA
RUO – Research Use Only