Learn More
3-Pyrrolidin-1-ylbenzoic acid, 97%, Thermo Scientific™
£49.55 - £92.00
Chemical Identifiers
CAS | 72548-79-9 |
---|---|
Molecular Formula | C11H13NO2 |
Molecular Weight (g/mol) | 191.23 |
MDL Number | MFCD03426724 |
InChI Key | HRVWGEKZONYEMK-UHFFFAOYSA-N |
PubChem CID | 651721 |
IUPAC Name | 3-pyrrolidin-1-ylbenzoic acid |
SMILES | C1CCN(C1)C2=CC=CC(=C2)C(=O)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10229822
|
Thermo Scientific Maybridge
CC38501CB |
250 mg | Amber Glass Bottle |
£49.55
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10621465
|
Thermo Scientific Maybridge
CC38501DA |
1 g | Amber Glass Bottle |
£92.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
72548-79-9 | |
191.23 | |
HRVWGEKZONYEMK-UHFFFAOYSA-N | |
3-pyrrolidin-1-ylbenzoic acid |
C11H13NO2 | |
MFCD03426724 | |
651721 | |
C1CCN(C1)C2=CC=CC(=C2)C(=O)O |
Specifications
72548-79-9 | |
100.0 | |
C11H13NO2 | |
3-pyrrolidin-1-yl benzoic acid, 3-pyrrolidin-1-yl-benzoic acid, 3-1-pyrrolidinyl benzoic acid, benzoic acid, 3-1-pyrrolidinyl, 3-pyrrolidinylbenzoic acid, pubchem19568, 3-pyrrolidinobenzoic acid, 3-1-pyrrolidinyl benzoicacid, 1-3-carboxyphenyl pyrrolidine, benzoic acid,3-1-pyrrolidinyl | |
C1CCN(C1)C2=CC=CC(=C2)C(=O)O | |
191.23 | |
191.23 | |
3-Pyrrolidin-1-ylbenzoic acid |
96.5 | |
97% | |
MFCD03426724 | |
HRVWGEKZONYEMK-UHFFFAOYSA-N | |
3-pyrrolidin-1-ylbenzoic acid | |
651721 | |
97% |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if inhaled.
Harmful if swallowed.
Causes skin irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Warning