Learn More
3-(Trifluoromethoxy)phenol, 98%, Thermo Scientific™
£23.50 - £135.45
Chemical Identifiers
CAS | 827-99-6 |
---|---|
Molecular Formula | C7H5F3O2 |
Molecular Weight (g/mol) | 178.11 |
MDL Number | MFCD00040987 |
InChI Key | UWLJERQTLRORJN-UHFFFAOYSA-N |
Synonym | 3-trifluoromethoxy phenol, m-trifluoromethoxy phenol, phenol, 3-trifluoromethoxy, 3-trifluoromethoxy-phenol, 3-hydroxyphenyl trifluoromethyl ether, pubchem1505, m-trifluoromethoxyphenol, pubchem10391 |
PubChem CID | 2733261 |
IUPAC Name | 3-(trifluoromethoxy)phenol |
SMILES | OC1=CC=CC(OC(F)(F)F)=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10183424
|
Thermo Scientific Acros
308702500 |
250 mg | Glass Bottle |
£23.50
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10115950
|
Thermo Scientific Acros
308700010 |
1 g | Glass bottle |
£43.60
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10091734
|
Thermo Scientific Acros
308700050 |
5 g | Glass bottle | ||||||
Description
Chemical Identifiers
827-99-6 | |
178.11 | |
UWLJERQTLRORJN-UHFFFAOYSA-N | |
2733261 | |
OC1=CC=CC(OC(F)(F)F)=C1 |
C7H5F3O2 | |
MFCD00040987 | |
3-trifluoromethoxy phenol, m-trifluoromethoxy phenol, phenol, 3-trifluoromethoxy, 3-trifluoromethoxy-phenol, 3-hydroxyphenyl trifluoromethyl ether, pubchem1505, m-trifluoromethoxyphenol, pubchem10391 | |
3-(trifluoromethoxy)phenol |
Specifications
827-99-6 | |
100.0 | |
1.3700g/mL | |
84°C | |
97.5% min. (GC) | |
CF3OC6H4OH | |
MFCD00040987 | |
3-trifluoromethoxy phenol, m-trifluoromethoxy phenol, phenol, 3-trifluoromethoxy, 3-trifluoromethoxy-phenol, 3-hydroxyphenyl trifluoromethyl ether, pubchem1505, m-trifluoromethoxyphenol, pubchem10391 | |
OC1=CC=CC(OC(F)(F)F)=C1 | |
178.11 | |
178.11 | |
Liquid |
97.5 | |
Colorless to Yellow | |
69.0°C to 70.0°C (12.0 mmHg) | |
Authentic | |
C7H5F3O2 | |
1.4450 to 1.4470 | |
1.37 | |
UWLJERQTLRORJN-UHFFFAOYSA-N | |
3-(trifluoromethoxy)phenol | |
2733261 | |
98% | |
3-(Trifluoromethoxy)phenol |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Harmful if inhaled.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
RUO – Research Use Only