Learn More
RETSCH 305 dia. x 40mmH Stainless Steel Test Sieve, ISO Certified, Micrometer Pore Sizes
Brand:  RETSCH 60.158.000075
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 10024791
  • £175.00 / Each
Estimated Shipment: 22-11-2024
to see stock.
Add to basket
Specifications
Stainless Steel
Stainless Steel
1Each
Test Sieve
Stainless Steel
Certificate indicating Compliance
For separation, fractioning, particle size determination
305 mm
75μm
ISO 3310/1
Shaker AS 300
40mm