Learn More
RETSCH 305 dia. x 40 mmH Stainless Steel Test Sieve, ISO Certified, Micrometer Pore Sizes
Brand:  RETSCH 60.158.000180
 View more versions of this product
Product Code. 10259211
  • £182.00 / Each
Estimated Shipment: 25-02-2025
to see stock.
Explore more special offers
Add to basket
Specifications
1Each
Test Sieve
Stainless Steel
Certificate indicating Compliance
For separation, fractioning, particle size determination
Stainless Steel
180 μm
ISO 3310/1
Shaker AS 300
40mm
305 mm
Stainless Steel