Learn More
RETSCH 305 dia. x 40mmH Stainless Steel Test Sieve, ISO Certified, Pore Sizes: Under 10mm
£175.00 - £216.00
Specifications
Diameter (Metric) Outer | 305 mm |
---|---|
Frame Material | Stainless Steel |
Mesh Material | Stainless Steel |
For Use With (Application) | For separation, fractioning, particle size determination |
Quantity | 1Each |
Product Code | Brand | Pore Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Pore Size | Price | Quantity & Availability | |||||
10085081
|
RETSCH
60.158.001120 |
1.12mm |
£175.00
Each |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||||
10711243
|
RETSCH
60.158.001180 |
1.18mm |
£175.00
Each |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||||
10711433
|
RETSCH
60.158.001400 |
1.40mm |
£175.00
Each |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||||
10584872
|
RETSCH
60.158.001250 |
1.25mm |
£175.00
Each |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||||
10219171
|
RETSCH
60.158.001600 |
1.60mm |
£175.00
Each |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||||
10487961
|
RETSCH
60.158.001800 |
1.80mm |
£175.00
Each |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||||
10074931
|
RETSCH
60.158.001700 |
1.70mm |
£175.00
Each |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||||
10310822
|
RETSCH
60.158.001000 |
1mm |
£175.00
Each |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||||
10034971
|
RETSCH
60.158.002240 |
2.24mm |
£175.00
Each |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||||
10739322
|
RETSCH
60.158.002360 |
2.36mm |
£213.00
Each |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||||
10370382
|
RETSCH
60.158.002500 |
2.50mm |
£213.00
Each |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||||
10218501
|
RETSCH
60.158.002800 |
2.80mm |
£212.00
Each |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||||
10116461
|
RETSCH
60.158.002000 |
2mm |
£216.00
Each |
Estimated Shipment: 02-10-2024
Log in to see stock. |
|||||
10710093
|
RETSCH
60.158.003150 |
3.15mm |
£212.00
Each |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||||
10478731
|
RETSCH
60.158.003350 |
3.35mm |
£212.00
Each |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||||
10478921
|
RETSCH
60.158.003550 |
3.55mm |
£212.00
Each |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||||
10156881
|
RETSCH
60.158.004500 |
4.50mm |
£212.00
Each |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||||
10593722
|
RETSCH
60.158.004750 |
4.75mm |
£212.00
Each |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||||
10166881
|
RETSCH
60.158.005600 |
5.60mm |
£212.00
Each |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||||
10720093
|
RETSCH
60.158.004000 |
4mm |
£212.00
Each |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||||
10075051
|
RETSCH
60.158.005000 |
5mm |
£212.00
Each |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||||
10054911
|
RETSCH
60.158.006300 |
6.30mm |
£212.00
Each |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||||
10157121
|
RETSCH
60.158.007100 |
7.10mm |
£212.00
Each |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||||
10630642
|
RETSCH
60.158.009000 |
9mm |
£212.00
Each |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||||
10004951
|
RETSCH
60.158.009500 |
9.50mm |
£212.00
Each |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||||
10064911
|
RETSCH
60.158.008000 |
8mm |
£212.00
Each |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||||
10486621
|
RETSCH
60.158.006700 |
6.70mm |
£212.00
Each |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||||
Spécification
305 mm | |
Stainless Steel | |
1Each | |
ISO 3310/1 | |
Shaker AS 300 | |
40mm |
Stainless Steel | |
For separation, fractioning, particle size determination | |
Test Sieve | |
Stainless Steel | |
Certificate indicating Compliance |