Learn More
Sampling Systems™ 316 Stainless Steel Storage Containers without Toggle Clamps
Multi-purpose containers with light duty construction for a wide range of storage uses in the laboratory.
Brand:  Sampling Systems™ A466-75
 View more versions of this product
Product Code. 15899681
  • £1520.00 / Each
Explore more special offers
Description
Description
  • Crevice-free interior
  • Two sturdy handles
  • Close fitting recessed lid with handle
  • Flat bottom with chime ring to give added protection
  • Toggle clamps and seals are optional extras
Specifications
Specifications
Yes
Recessed Lid
Stainless Steel
400 mm
Storage container
75 L
625 mm