missing translation for 'onlineSavingsMsg'
Learn More

Nalgene 32 oz. Wide Mouth Bottle

The Nalgene 32 oz. Wide Mouth water bottle is made with durable Tritan plastic, BPA/BPS-free, and made in the USA.

Brand:  Nalgene 2178-2040

Additional Details : Weight : 1.29000kg

 View more versions of this product

Product Code. 17100066

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

  • Expertly crafted threads for leak-proof hydration
  • Wide mouth that's ideal for ice cubes or fruit infusions
  • Proven in the elements and ready to withstand even the most rugged conditions
Specifications

Specifications

946 mL
Trout Green
Wide
32 oz.
Plastic
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers