missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nalgene 32 oz. Wide Mouth Bottle
The Nalgene 32 oz. Wide Mouth water bottle is made with durable Tritan plastic, BPA/BPS-free, and made in the USA.
Brand: Nalgene 2178-2053
Additional Details : Weight : 1.26000kg
Description
- Expertly crafted threads for leak-proof hydration
- Wide mouth that's ideal for ice cubes or fruit infusions
- Proven in the elements and ready to withstand even the most rugged conditions
Specifications
946 mL | |
Seafoam | |
Wide |
32 oz. | |
Plastic |