missing translation for 'onlineSavingsMsg'
Learn More

Nalgene 32 oz. Wide Mouth Bottle

The Nalgene 32 oz. Wide Mouth water bottle is made with durable Tritan plastic, BPA/BPS-free, and made in the USA.

Brand:  Nalgene 2178-2025

Additional Details : Weight : 2.47000kg

 View more versions of this product

Product Code. 17190056

  • £11.85 / Each
In Stock
to see stock.
Add to basket

You may only order up to the available stock. Where an alternative item has been identified it will be shown below

Description

Description

  • Expertly crafted threads for leak-proof hydration
  • Wide mouth that's ideal for ice cubes or fruit infusions
  • Proven in the elements and ready to withstand even the most rugged conditions
Specifications

Specifications

946 mL
Gray
Wide
32 oz.
Plastic
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers