Learn More
Brannan™ ECO-therm Lab Thermometer
Green spirit thermometers filled with non toxic expansion liquid.
Brand:  Brannan™ 46/706/0
 View more versions of this product
Product Code. 15472685
  • £30.10 / Pack of 10
Estimated Shipment: 11-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Quality and resolution make these ideal substitute for general purpose mercury thermometers
  • Each thermometer is fitted with Brannan non roll end cap
  • Cap has integral ring to allow the thermometer to be suspended and is supplied in protective PVC packaging
Specifications
Specifications
Partial Immersion
Glass
Laboratory Immersion Thermometer
-10°C
±1.5°C
305 mm
Green Spirit
150°C
-10°C to +150°C
1°C