Learn More
Corning™ 384 Well Flat Clear Bottom Polystyrene Untreated Microplates, low flange, no lid, nonsterile
Untreated (medium binding) polystyrene surface is hydrophobic in nature and binds biomolecules through passive interactions. Corning™ 384 Well Flat Clear Bottom Polystyrene Untreated Microplates are ideally suited for fluorescent assays using either top or bottom detection plate readers.
Brand: Corning™ 3762
Description
- Recommended working volume of up to 20 to 80μL
- Conforms to standard microplate footprint and dimensions
- Not treated (or medium binding) polystyrene surface is hydrophobic in nature and binds biomolecules through passive interactions
- Suitable primarily for the immobilization of large molecules, such as antibodies, that have large hydrophobic regions that can interact with the surface
- Untreated microplates have a binding capacity of approximately 100 to 200ng lgG/cm²
Specifications
Standard | |
Black | |
384 | |
Non-sterile | |
Square, Flat Bottom (F-well) |
16 x 24 | |
Polystyrene | |
20 to 80 μL | |
Non-treated |