missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Titanium(III) chloride, 20% w/v solution in 2N Hydrochloric acid, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 395620010
289.00 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
Specifications
Titanium(III) chloride | |
Purple | |
1 L | |
TiCl3 | |
15,9637 | |
Solubility in water: soluble. Other solubilities: soluble in alcohol, practically insoluble in ether | |
[Cl-].[Cl-].[Cl-].[Ti+3] | |
154.22 | |
∼20% (TiCl3) (exact strength stated on certificate of analysis) | |
1.2000g/mL |
7705-07-9, 7647-01-0, 7732-18-5 | |
Liquid | |
Cl3Ti | |
MFCD00011266 | |
Titanous chloride | |
YONPGGFAJWQGJC-UHFFFAOYSA-K | |
titanium(3+) trichloride | |
154.26 | |
Plastic bottle |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Titanium(III) chloride, 20% w/v solution in 2N Hydrochloric acid, Thermo Scientific Chemicals > 1L; Plastic bottle
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction