missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ 384-Well Polypropylene Plates
GREENER_CHOICE

Feature higher well density for economical use of expensive reagents

Brand:  Fisherbrand™ 235800

337.25 GBP valid until 2025-04-30
Use promo code "24163" to get your promotional price.


 View more versions of this product

Product Code. 11957954

  • £355.00 / Pack of 120
Estimated Shipment: 07-08-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Ideal for storage of compounds and nucleic acid libraries
  • Chemical-resistant
  • Withstand temperatures from -80° to +80°C (-112° to +176°F)
  • Conform to ANSI™-standard footprint for optimal use in automated handling and liquid-dispensing systems

TRUSTED_SUSTAINABILITY
Specifications

Specifications

ANSI
16 x 24
384-Well Storage Microplate
384
For storage of compounds and nucleic acid libraries
Standard
Natural
Polypropylene
Non-sterile
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisherbrand™ 384-Well Polypropylene Plates > Natural; Well vol.: 120μL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.