Learn More
3M™ TR-300 and TR-600 PAPR Battery Packs
Choose from a selection of lithium ion rechargeable batteries for the TR-300 or TR-600 PAPR systems.
Brand:  3M™ 7000144502
317.73 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 15589255
Description
Description
  • Rechargeable battery packs
  • Lithium Ion
  • Long run time for filters and cartridges
Battery packs must be used in accordance with the user instructions, warnings and limitations accompanying each product
Compliance
NIOSH approved.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
8 to 18 hr.
Blue, Gray
Supplying power for PAPR systems
TR-632
Lithium-ion
Battery Pack
Versaflo TR-600 PAPR Series
1/Cs.
Product Suggestions
Product Suggestions
Videos
Videos
  • 3M Versaflo powered air turbo TR-600
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
3M™ TR-300 and TR-600 PAPR Battery Packs > Model TR-632, 8-18 hours, Versaflo, Blue/Gray, Lithium Ion, Rechargeable, TR-600 series PAPR