Learn More
3M™ Versaflo™ Battery TR-830/94243(AAD)
3M™ Versaflo™ Battery TR-830/94243(AAD) is a lithium-ion battery pack designed for use with the 3M™ Versaflo™ TR-800 Series Powered Air Turbo.
Brand:  3M™ 7100150925
587.44 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 16446360
  • £880.00 / Each
Explore more special offers
Description
Description
  • Charges quickly (90 % charge in approximately 3.5 hours)
  • Approximately 750 full charge/discharge cycles
  • Standard capacity rechargeable lithium ion battery
  • Mechanical attachment of battery to help prevent battery removal and spark risk during use per EN 60079-11
  • Built in charge level indicator
  • Intrinsically safe
TRUSTED_SUSTAINABILITY
Specifications
Specifications
6 to 10 hr
CE Approved, NIOSH Approved
TR-830
Lithium-ion
Battery
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
3M™ Versaflo™ Battery TR-830/94243(AAD) > Battery Type: Lithium Ion Rechargeable