Learn More
3M™ Standard Chemical Sorbents
Absorb virtually any liquid with these chemical sorbents
£335.22 - £1080.00
Specifications
Certifications/Compliance | OSHA 29 CFR 1910.132 |
---|---|
Color | Yellow |
Material | Polypropylene |
Product Code | Brand | Model | Capacity (Metric) | Capacity (English) | Type | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Model | Capacity (Metric) | Capacity (English) | Type | Price | Quantity & Availability | ||||||
12342459
|
3M™
7000051994 |
P200 | 45.4 L | 12 gal. | Chemical Sorbent Mini Boom |
£550.00
Pack of 12 |
Estimated Shipment: 12-05-2025
Log in to see stock. |
||||||
10685315
|
3M™
7000088220 |
P110 | 64.3 L | 17 gal. | Chemical Sorbent Pad |
£1080.00
Pack of 200 |
In Stock
Log in to see stock. ![]() |
||||||
12332459
|
3M™
7000052037 |
P300 | 30.3 L | 8 gal. | Chemical Sorbent Pillow |
£515.00
Pack of 16 |
Estimated Shipment: 10-06-2025
Log in to see stock. |
||||||
12362459
|
3M™
P-500 |
P500 | 53 L | 14 gal. | Chemical Sorbent Particulate | N/A | N/A | N/A | |||||
Description
Meltblown polypropylene microfibers absorb liquids such as hydrofluoric acid and other aqueous-based chemicals, and soak up an average of 10 times their weight of most EPA-designated hazardous liquids.
- Sorbents are dirt- and dust-free; contain no abrasive granules that can damage expensive equipment
- Polypropylene base is chemically inert and incinerable; allows separation and recycling in many applications
- Easy-to-spot yellow cartons protect sorbents from temperature and humidity for secure, long-term storage
- Convenient dispenser boxes—suitable for tabletops or wall mounting—keep pads and pillows within easy reach Pads or rolls are ideal for use on work surfaces and can be trimmed to size
- Mini booms surround and contain spills; pillows soak them up
- 3M Catalog# P110, P200, P208, P300, P500
These safety products must be used in accordance with the user instructions, warnings and limitations accompanying each product.
Specifications
OSHA 29 CFR 1910.132 | |
Yellow | |
Polypropylene |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.