missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(2-Methyl-3-furylthio)-5-nonanone, 95%, Thermo Scientific™
£49.35 - £134.00
Chemical Identifiers
CAS | 61295-50-9 |
---|---|
Molecular Formula | C14H22O2S |
Molecular Weight (g/mol) | 254.388 |
MDL Number | MFCD08741453 |
InChI Key | PEYZZTQOVLTVHN-UHFFFAOYSA-N |
Synonym | 4-2-methyl-3-furyl thio-5-nonanone, 4-2-methyl-3-furyl thio nonan-5-one, 4-2-methyl-3-furanyl thio-5-nonanone, 1,3-dipropylacetonyl 2-methyl-3-furyl sulfide, 4-2-methyl-furan-3-ylsulfanyl-nonan-5-one, 4-2-methyl-3-furylthio-5-nonanone, 4-2-methylfuran-3-yl sulfanylnonan-5-one, 5-nonanone, 4-2-methyl-3-furanyl thio, 4-2-methylfuran-3-yl sulfanyl nonan-5-one |
PubChem CID | 62182 |
IUPAC Name | 4-(2-methylfuran-3-yl)sulfanylnonan-5-one |
SMILES | CCCCC(=O)C(CCC)SC1=C(OC=C1)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11461860
|
Thermo Scientific Alfa Aesar
H27560.MC |
100 mg |
£49.35
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11471860
|
Thermo Scientific Alfa Aesar
H27560.ME |
500 mg | N/A | ||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
61295-50-9 | |
254.388 | |
PEYZZTQOVLTVHN-UHFFFAOYSA-N | |
62182 | |
CCCCC(=O)C(CCC)SC1=C(OC=C1)C |
C14H22O2S | |
MFCD08741453 | |
4-2-methyl-3-furyl thio-5-nonanone, 4-2-methyl-3-furyl thio nonan-5-one, 4-2-methyl-3-furanyl thio-5-nonanone, 1,3-dipropylacetonyl 2-methyl-3-furyl sulfide, 4-2-methyl-furan-3-ylsulfanyl-nonan-5-one, 4-2-methyl-3-furylthio-5-nonanone, 4-2-methylfuran-3-yl sulfanylnonan-5-one, 5-nonanone, 4-2-methyl-3-furanyl thio, 4-2-methylfuran-3-yl sulfanyl nonan-5-one | |
4-(2-methylfuran-3-yl)sulfanylnonan-5-one |
Specifications
61295-50-9 | |
1.4985 | |
4-2-methyl-3-furyl thio-5-nonanone, 4-2-methyl-3-furyl thio nonan-5-one, 4-2-methyl-3-furanyl thio-5-nonanone, 1,3-dipropylacetonyl 2-methyl-3-furyl sulfide, 4-2-methyl-furan-3-ylsulfanyl-nonan-5-one, 4-2-methyl-3-furylthio-5-nonanone, 4-2-methylfuran-3-yl sulfanylnonan-5-one, 5-nonanone, 4-2-methyl-3-furanyl thio, 4-2-methylfuran-3-yl sulfanyl nonan-5-one | |
CCCCC(=O)C(CCC)SC1=C(OC=C1)C | |
254.388 | |
254.39 | |
4-(2-Methyl-3-furylthio)-5-nonanone |
C14H22O2S | |
MFCD08741453 | |
PEYZZTQOVLTVHN-UHFFFAOYSA-N | |
4-(2-methylfuran-3-yl)sulfanylnonan-5-one | |
62182 | |
95% |
Safety and Handling
missing translation for 'einecsNumber' : 262-691-7
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures