missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(3-Chloro-4-methylphenylcarbamoyl)benzeneboronic acid, 98%, Thermo Scientific™
£44.50 - £138.00
Chemical Identifiers
CAS | 913835-37-7 |
---|---|
Molecular Formula | C14H13BClNO3 |
Molecular Weight (g/mol) | 289.522 |
MDL Number | MFCD09027211 |
InChI Key | IZMAVYDCXUROFV-UHFFFAOYSA-N |
Synonym | n-3-chloro-4-methylphenyl 4-boronobenzamide, 4-3-chloro-4-methylphenyl carbamoyl phenyl boronic acid, 4-3-chloro-4-methylphenyl carbamoyl benzeneboronic acid, 4-3-chloro-4-methylphenyl carbamoyl phenylboronic acid, 4-3-chloro-4-methylphenyl carbamoyl phenyl boronicacid, 4-3-chloro-4-methylphenyl carbamoyl-phenyl boronic acid, boronic acid,b-4-3-chloro-4-methylphenyl amino carbonyl phenyl |
PubChem CID | 44119159 |
IUPAC Name | [4-[(3-chloro-4-methylphenyl)carbamoyl]phenyl]boronic acid |
SMILES | B(C1=CC=C(C=C1)C(=O)NC2=CC(=C(C=C2)C)Cl)(O)O |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
913835-37-7 | |
289.522 | |
IZMAVYDCXUROFV-UHFFFAOYSA-N | |
44119159 | |
B(C1=CC=C(C=C1)C(=O)NC2=CC(=C(C=C2)C)Cl)(O)O |
C14H13BClNO3 | |
MFCD09027211 | |
n-3-chloro-4-methylphenyl 4-boronobenzamide, 4-3-chloro-4-methylphenyl carbamoyl phenyl boronic acid, 4-3-chloro-4-methylphenyl carbamoyl benzeneboronic acid, 4-3-chloro-4-methylphenyl carbamoyl phenylboronic acid, 4-3-chloro-4-methylphenyl carbamoyl phenyl boronicacid, 4-3-chloro-4-methylphenyl carbamoyl-phenyl boronic acid, boronic acid,b-4-3-chloro-4-methylphenyl amino carbonyl phenyl | |
[4-[(3-chloro-4-methylphenyl)carbamoyl]phenyl]boronic acid |
Specifications
913835-37-7 | |
C14H13BClNO3 | |
n-3-chloro-4-methylphenyl 4-boronobenzamide, 4-3-chloro-4-methylphenyl carbamoyl phenyl boronic acid, 4-3-chloro-4-methylphenyl carbamoyl benzeneboronic acid, 4-3-chloro-4-methylphenyl carbamoyl phenylboronic acid, 4-3-chloro-4-methylphenyl carbamoyl phenyl boronicacid, 4-3-chloro-4-methylphenyl carbamoyl-phenyl boronic acid, boronic acid,b-4-3-chloro-4-methylphenyl amino carbonyl phenyl | |
B(C1=CC=C(C=C1)C(=O)NC2=CC(=C(C=C2)C)Cl)(O)O | |
289.522 | |
289.53 | |
4-(3-Chloro-4-methylphenylcarbamoyl)benzeneboronic acid |
217°C to 219°C | |
MFCD09027211 | |
IZMAVYDCXUROFV-UHFFFAOYSA-N | |
[4-[(3-chloro-4-methylphenyl)carbamoyl]phenyl]boronic acid | |
44119159 | |
98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures