missing translation for 'onlineSavingsMsg'
Learn More

4-(3-Chlorophenoxy)piperidine, 98%, Thermo Scientific Chemicals

Product Code. 11409710
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 11409710

Brand: Thermo Scientific Alfa Aesar H50956.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97840-40-9
Molecular Formula C11H14ClNO
Molecular Weight (g/mol) 211.689
MDL Number MFCD08061087
InChI Key BYXWGMQMUJCYPK-UHFFFAOYSA-N
Synonym 4-3-chlorophenoxy piperidine, 4-3-chloro-phenoxy-piperidine, piperidine, 4-3-chlorophenoxy, acmc-20amww, 4-m-chlorophenoxypiperidine, 4-3-chlorophenoxy-piperidine, piperidine,4-3-chlorophenoxy, 1-chloro-3-4-piperidyloxy benzene, piperidine, 4-3-chlorophenoxy-, hydrochloride
PubChem CID 2735317
IUPAC Name 4-(3-chlorophenoxy)piperidine
SMILES C1CNCCC1OC2=CC(=CC=C2)Cl

Specifications

Quantity 1 g
Sensitivity Air Sensitive
Formula Weight 211.69
Percent Purity 98%
Chemical Name or Material 4-(3-Chlorophenoxy)piperidine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.