accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 97840-40-9

CAS RN 97840-40-9

IUPAC Name: 4-(3-chlorophenoxy)piperidine
Synonyms: 4-(3-chlorophenoxy)piperidine
Molecular Weight (g/mol): 211.69
Molecular Formula: C11H14ClNO
InChi Key: BYXWGMQMUJCYPK-UHFFFAOYSA-N
SMILES: ClC1=CC=CC(OC2CCNCC2)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.