Learn More
Nabertherm™ Chimney
Chimney with fan, to remove exhaust gas from the furnace better
Brand: Nabertherm™ 6000140311
This item is not returnable.
View return policy
Description
not suitable for
- L 15...
- L 1/12
- LE 1/11
- LE 2/12
Specifications
P330 | |
Chimney | |
To Remove Exhaust Gas From The Furnace Better | |
Fan |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Product Title
Nabertherm™ Chimney > For Use With: To remove exhaust gas from the furnace better
Spot an opportunity for improvement?Share a Content Correction