Learn More
Nabertherm Catalytic Converter with Fan
For removal of organic components from the exhaust air.
Brand:  Nabertherm 631000166
Product Code. 11990098
  • £1095.00 / Each
Estimated Shipment: 22-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • For the removal of organic components from the exhaust air.
  • Organic components are catalytically oxidized at approximately 600°C, broken into carbon dioxide and water vapor.
  • Irritating odors are largely eliminated.
  • CE Marked
Specifications
Specifications
Removal of organic components from the exhaust air
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Nabertherm Catalytic Converter with Fan > For use with Nabertherm Burnout Furnaces