Learn More
Memmert™ Full-Sight Glass Door
Memmert™ Full-Sight Glass Door with 5-layer insulating glazing for use with Environmental Test Chambers.
Brand:  Memmert™ CTC256B0
Product Code. 15745288
  • £1290.00 / Each
Estimated Shipment: 14-07-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • 5-layer insulating glazing
  • Heated
Specifications
Specifications
CTC256
Accessory, Full-Sight Door
5-layer insulating glazing
Glass
Full-sight
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Memmert™ Full-Sight Glass Door > For Use With: CTC256