Learn More
Invitrogen™ TBE (10X), RNase-free
Molecular biology grade, Invitrogen 10X TBE solution is supplied in four bottles containing 1 L each. The buffer is certified RNase-free, economical, and ready-to-use.
Brand: Invitrogen™ AM9865
140.64 GBP valid until 2025-07-13
Use promo code "24135" to get your promotional price.
Description
Molecular biology grade, Invitrogen 10X TBE solution is supplied in four bottles containing 1 L each. The buffer is certified RNase-free, economical, and ready-to-use. Due to the ubiquitous presence of RNases, manufacturing products for use with RNA is especially challenging. Invitrogen's nuclease-free reagents and buffers are manufactured in facilities specifically designed to prevent the introduction of nucleases. Highly sensitive RNase assays are performed at several different stages of the manufacturing process to ensure the highest quality. These reagents are rigorously tested for contaminating nonspecific endonuclease, exonuclease, and RNase activity.
Order Info
Shipping Condition: Room temperature
Certifications
Certified RNase-free

Specifications
Running Buffer | |
Liquid | |
4 x 1 L |
10X | |
Ambion | |
Molecular Biology |
Safety and Handling
- TBE (10X), RNase-free
Signal Word
- Danger
Hazard Category
- Reproductive toxicity Category 1B
Hazard Statement
- H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Boric acid (H3BO3)
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Special Offers
-
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
New Offer Save Up to 85%
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.