missing translation for 'onlineSavingsMsg'
Learn More
4,4',4″-Trimethyltriphenylamine, 98%, Thermo Scientific Chemicals Product Code.: 15411239

4,4',4″-Trimethyltriphenylamine, 98%, Thermo Scientific Chemicals

Product Code. 15411239
1 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 15411239

Brand: Thermo Scientific Alfa Aesar H64162.03

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used for proteomics research. Also employed as a intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used for proteomics research. Also employed as a intermediate for pharmaceutical.

Solubility
Soluble in chloroform and tetrahydrofuran (THF).

Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1159-53-1
Molecular Formula C21H21N
Molecular Weight (g/mol) 287.406
MDL Number MFCD00674043
InChI Key YXYUIABODWXVIK-UHFFFAOYSA-N
Synonym tri-p-tolylamine, 4,4',4-trimethyltriphenylamine, 4,4,4-trimethyltriphenylamine, tri p-tolyl amine, benzenamine, 4-methyl-n,n-bis 4-methylphenyl, 4-methyl-n,n-bis 4-methylphenyl aniline, tris 4-methylphenyl amine, n,n-di-p-tolyl-p-toluidine, n,n,n-tri p-tolyl amine, trip-tolylamineShow MoreShow Less
PubChem CID 70873
IUPAC Name 4-methyl-N,N-bis(4-methylphenyl)aniline
SMILES CC1=CC=C(C=C1)N(C2=CC=C(C=C2)C)C3=CC=C(C=C3)C

Specifications

Melting Point 114°C to 118°C (literature)
Color White
Quantity 1 g
Beilstein 2215309
Solubility Information Soluble in chloroform and tetrahydrofuran (THF).
Formula Weight 287.41
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 4,4',4''-Trimethyltriphenylamine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4,4',4″-Trimethyltriphenylamine, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.