missing translation for 'onlineSavingsMsg'
Learn More

4-(4-bromo-1,3-thiazol-2-yl)morpholine, 97%, Thermo Scientific™

£48.83 - £172.47

Chemical Identifiers

CAS 1017781-60-0
Molecular Formula C7H9BrN2OS
Molecular Weight (g/mol) 249.126
MDL Number MFCD09607696
InChI Key XPFAGXZWVZAYCS-UHFFFAOYSA-N
Synonym 4-4-bromothiazol-2-yl morpholine, 4-4-bromo-1,3-thiazol-2-yl morpholine, morpholine, 4-4-bromo-2-thiazolyl, 4-4-bromo-thiazol-2-yl-morpholine, acmc-2097ww, 4-bromo-2-morpholinothiazole, morpholine,4-4-bromo-2-thiazolyl, 4-bromo-2-morpholin-4-yl-1,3-thiazole
PubChem CID 43811042
IUPAC Name 4-(4-bromo-1,3-thiazol-2-yl)morpholine
SMILES C1COCCN1C2=NC(=CS2)Br
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10758655
View Documents
Thermo Scientific Maybridge
CC62810DA
1 g Amber Glass Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10295574
View Documents
Thermo Scientific Maybridge
CC62810DE
5 g Amber Glass Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Specifications

Chemical Identifiers

1017781-60-0
249.126
XPFAGXZWVZAYCS-UHFFFAOYSA-N
43811042
C1COCCN1C2=NC(=CS2)Br
C7H9BrN2OS
MFCD09607696
4-4-bromothiazol-2-yl morpholine, 4-4-bromo-1,3-thiazol-2-yl morpholine, morpholine, 4-4-bromo-2-thiazolyl, 4-4-bromo-thiazol-2-yl-morpholine, acmc-2097ww, 4-bromo-2-morpholinothiazole, morpholine,4-4-bromo-2-thiazolyl, 4-bromo-2-morpholin-4-yl-1,3-thiazole
4-(4-bromo-1,3-thiazol-2-yl)morpholine

Specifications

1017781-60-0
100.0
C7H9BrN2OS
4-4-bromothiazol-2-yl morpholine, 4-4-bromo-1,3-thiazol-2-yl morpholine, morpholine, 4-4-bromo-2-thiazolyl, 4-4-bromo-thiazol-2-yl-morpholine, acmc-2097ww, 4-bromo-2-morpholinothiazole, morpholine,4-4-bromo-2-thiazolyl, 4-bromo-2-morpholin-4-yl-1,3-thiazole
C1COCCN1C2=NC(=CS2)Br
249.126
249.13
4-(4-Bromo-1, 3-thiazol-2-yl)morpholine
96.5
97%
MFCD09607696
XPFAGXZWVZAYCS-UHFFFAOYSA-N
4-(4-bromo-1,3-thiazol-2-yl)morpholine
43811042
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents

Product Certifications