missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dichlorobutyrophenone, 97%, Thermo Scientific™

£31.00 - £85.30

Chemical Identifiers

CAS 40877-09-6
Molecular Formula C10H10Cl2O
Molecular Weight (g/mol) 217.09
MDL Number MFCD00018993
InChI Key UKCHLVFIVJBCKE-UHFFFAOYSA-N
Synonym 4-chloro-1-4-chlorophenyl butan-1-one, 4,4'-dichlorobutyrophenone, 1-butanone, 4-chloro-1-4-chlorophenyl, gamma,p-dichlorobutyrophenone, 4-chloro-1-4-chlorophenyl-1-oxobutane, 4-chloro-1-4-chlorophenyl-1-butanone, acmc-20a5su, 4',gamma-dichlorobutyrophenone, 4-chloro-4'-chlorobutyrophenone, 4-chloro-4'-chloro-butyrophenone
PubChem CID 2723781
IUPAC Name 4-chloro-1-(4-chlorophenyl)butan-1-one
SMILES C1=CC(=CC=C1C(=O)CCCCl)Cl
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10396490
View Documents Promotion Details
Thermo Scientific Acros
136890250
25 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
10193660
View Documents Promotion Details
Thermo Scientific Acros
136891000
100 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Specifications

Chemical Identifiers

40877-09-6
217.09
UKCHLVFIVJBCKE-UHFFFAOYSA-N
2723781
C1=CC(=CC=C1C(=O)CCCCl)Cl
C10H10Cl2O
MFCD00018993
4-chloro-1-4-chlorophenyl butan-1-one, 4,4'-dichlorobutyrophenone, 1-butanone, 4-chloro-1-4-chlorophenyl, gamma,p-dichlorobutyrophenone, 4-chloro-1-4-chlorophenyl-1-oxobutane, 4-chloro-1-4-chlorophenyl-1-butanone, acmc-20a5su, 4',gamma-dichlorobutyrophenone, 4-chloro-4'-chlorobutyrophenone, 4-chloro-4'-chloro-butyrophenone
4-chloro-1-(4-chlorophenyl)butan-1-one

Specifications

40877-09-6
100.0
White
96% min. (GC)
C10H10Cl2O
MFCD00018993
Solubility in water: insoluble
C1=CC(=CC=C1C(=O)CCCCl)Cl
217.09
217.09
Pellets
96.0
29°C to 30°C
Authentic
Glass Bottle
ClC6H4CO(CH2)3Cl
4-chloro-1-4-chlorophenyl butan-1-one, 4,4'-dichlorobutyrophenone, 1-butanone, 4-chloro-1-4-chlorophenyl, gamma,p-dichlorobutyrophenone, 4-chloro-1-4-chlorophenyl-1-oxobutane, 4-chloro-1-4-chlorophenyl-1-butanone, acmc-20a5su, 4',gamma-dichlorobutyrophenone, 4-chloro-4'-chlorobutyrophenone, 4-chloro-4'-chloro-butyrophenone
UKCHLVFIVJBCKE-UHFFFAOYSA-N
4-chloro-1-(4-chlorophenyl)butan-1-one
2723781
97%
4, 4'-Dichlorobutyrophenone, 97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 255-123-4

missing translation for 'tsca' : TSCA

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only