missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(5-bromothien-2-yl)pyridine, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Specifications
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Linear Formula | Yes |
Quantity | 5 g |
Formula Weight | 267.08 |
Percent Purity | 97% |
Chemical Name or Material | 4-(5-Bromothien-2-yl)pyridine |
Product Suggestions
Customers who viewed this item also viewed
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
4-(5-bromothien-2-yl)pyridine, 97%, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction