Learn More
4,7-Dimethyl-4-octanol, 99%, Thermo Scientific™
£15.70 - £204.00
Chemical Identifiers
CAS | 19781-13-6 |
---|---|
Molecular Formula | C10H22O |
Molecular Weight (g/mol) | 158.285 |
MDL Number | MFCD00046603 |
InChI Key | OTISWHNJCUMBFI-UHFFFAOYSA-N |
PubChem CID | 89224 |
IUPAC Name | 4,7-dimethyloctan-4-ol |
SMILES | CCCC(C)(CCC(C)C)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11449686
|
Thermo Scientific Alfa Aesar
B20225.03 |
1 g |
£15.70
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11459686
|
Thermo Scientific Alfa Aesar
B20225.06 |
5 g | N/A | ||||||
11469686
|
Thermo Scientific Alfa Aesar
B20225.14 |
25 g | N/A | ||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
19781-13-6 | |
158.285 | |
OTISWHNJCUMBFI-UHFFFAOYSA-N | |
4,7-dimethyloctan-4-ol |
C10H22O | |
MFCD00046603 | |
89224 | |
CCCC(C)(CCC(C)C)O |
Specifications
19781-13-6 | |
MFCD00046603 | |
OTISWHNJCUMBFI-UHFFFAOYSA-N | |
4,7-dimethyloctan-4-ol | |
89224 | |
99% |
C10H22O | |
4,7-dimethyl-4-octanol, 4-octanol, 4,7-dimethyl, 2,5-dimethyl-5-octanol, acmc-20amr9, 2.5-dimethyloctanol-5, 4-octanol,4,7-dimethyl | |
CCCC(C)(CCC(C)C)O | |
158.285 | |
158.29 | |
4,7-Dimethyl-4-octanol |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 243-306-1
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures