Learn More
4-Acetamidophenylboronic acid, 97+%, Thermo Scientific Chemicals
£72.30 - £249.00
Chemical Identifiers
CAS | 101251-09-6 |
---|---|
Molecular Formula | C8H10BNO3 |
Molecular Weight (g/mol) | 178.98 |
MDL Number | MFCD02179451 |
InChI Key | VYEWTHXZHHATTA-UHFFFAOYSA-N |
Synonym | 4-acetylaminophenylboronic acid, 4-acetamidophenyl boronic acid, p-acetamidophenylboronic acid, 4-acetamidobenzeneboronic acid, 4-acetamidophenyl-boronic acid, 4-acetaminophenyl boronic acid, 4-acetylaminophenyl boronic acid, 4-acetylamino phenylboronic acid, boronic acid, 4-acetylamino phenyl |
PubChem CID | 2734657 |
IUPAC Name | (4-acetamidophenyl)boronic acid |
SMILES | CC(=O)NC1=CC=C(C=C1)B(O)O |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
101251-09-6 | |
178.98 | |
VYEWTHXZHHATTA-UHFFFAOYSA-N | |
2734657 | |
CC(=O)NC1=CC=C(C=C1)B(O)O |
C8H10BNO3 | |
MFCD02179451 | |
4-acetylaminophenylboronic acid, 4-acetamidophenyl boronic acid, p-acetamidophenylboronic acid, 4-acetamidobenzeneboronic acid, 4-acetamidophenyl-boronic acid, 4-acetaminophenyl boronic acid, 4-acetylaminophenyl boronic acid, 4-acetylamino phenylboronic acid, boronic acid, 4-acetylamino phenyl | |
(4-acetamidophenyl)boronic acid |
Specifications
101251-09-6 | |
100.0 | |
Gray-Tan | |
97% min. (GC) | |
MFCD02179451 | |
Solubility in water: 85g/L (70°C). Other solubilities: slightly soluble in ethylacetate,soluble in meoh,etoh,dmso | |
CC(=O)NC1=CC=C(C=C1)B(O)O | |
178.98 | |
178.98 | |
Powder |
97.0 | |
215.0°C to 220.0°C | |
Authentic | |
C8H10BNO3 | |
4-acetylaminophenylboronic acid, 4-acetamidophenyl boronic acid, p-acetamidophenylboronic acid, 4-acetamidobenzeneboronic acid, 4-acetamidophenyl-boronic acid, 4-acetaminophenyl boronic acid, 4-acetylaminophenyl boronic acid, 4-acetylamino phenylboronic acid, boronic acid, 4-acetylamino phenyl | |
VYEWTHXZHHATTA-UHFFFAOYSA-N | |
(4-acetamidophenyl)boronic acid | |
2734657 | |
97+% | |
4-Acetamidophenylboronic acid |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
Recommended Storage : Refrigerator +4°C
- 4-Acetamidophenylboronic acid
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only