Learn More
4-(Benzyloxy)-2-fluorophenylboronic acid, 97%, anhydride present, Thermo Scientific™
£17.35 - £47.20
Chemical Identifiers
CAS | 166744-78-1 |
---|---|
Molecular Formula | C13H12BFO3 |
Molecular Weight (g/mol) | 246.044 |
MDL Number | MFCD02093064 |
InChI Key | PQWKGFALWCOVTC-UHFFFAOYSA-N |
Synonym | 4-benzyloxy-2-fluorophenylboronic acid, 4-benzyloxy-2-fluorobenzeneboronic acid, 4-benzyloxy-2-fluorophenyl boronic acid, boronic acid, 2-fluoro-4-phenylmethoxy phenyl, 2-fluoro-4-phenylmethoxyphenyl boronic acid, 4-benzyloxy-2-fluorophenylboronic acid, may contain varying amounts of anhydride, pubchem7760, akos brn-0077, acmc-209dv0 |
PubChem CID | 2737786 |
IUPAC Name | (2-fluoro-4-phenylmethoxyphenyl)boronic acid |
SMILES | B(C1=C(C=C(C=C1)OCC2=CC=CC=C2)F)(O)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10782683
|
Thermo Scientific Maybridge
MO00843CB |
250 mg | Amber Glass Bottle |
£17.35
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10600923
|
Thermo Scientific Maybridge
MO00843DA |
1 g | Amber Glass Bottle |
£47.20
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
166744-78-1 | |
246.044 | |
PQWKGFALWCOVTC-UHFFFAOYSA-N | |
2737786 | |
B(C1=C(C=C(C=C1)OCC2=CC=CC=C2)F)(O)O |
C13H12BFO3 | |
MFCD02093064 | |
4-benzyloxy-2-fluorophenylboronic acid, 4-benzyloxy-2-fluorobenzeneboronic acid, 4-benzyloxy-2-fluorophenyl boronic acid, boronic acid, 2-fluoro-4-phenylmethoxy phenyl, 2-fluoro-4-phenylmethoxyphenyl boronic acid, 4-benzyloxy-2-fluorophenylboronic acid, may contain varying amounts of anhydride, pubchem7760, akos brn-0077, acmc-209dv0 | |
(2-fluoro-4-phenylmethoxyphenyl)boronic acid |
Specifications
166744-78-1 | |
97% | |
MFCD02093064 | |
PQWKGFALWCOVTC-UHFFFAOYSA-N | |
(2-fluoro-4-phenylmethoxyphenyl)boronic acid | |
2737786 | |
97% |
97.0 | |
C13H12BFO3 | |
4-benzyloxy-2-fluorophenylboronic acid, 4-benzyloxy-2-fluorobenzeneboronic acid, 4-benzyloxy-2-fluorophenyl boronic acid, boronic acid, 2-fluoro-4-phenylmethoxy phenyl, 2-fluoro-4-phenylmethoxyphenyl boronic acid, 4-benzyloxy-2-fluorophenylboronic acid, may contain varying amounts of anhydride, pubchem7760, akos brn-0077, acmc-209dv0 | |
B(C1=C(C=C(C=C1)OCC2=CC=CC=C2)F)(O)O | |
246.044 | |
246.04 | |
4-(Benzyloxy)-2-fluorophenylboronic acid, May contain varying amounts of anhydride |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.