missing translation for 'onlineSavingsMsg'
Learn More

4-Benzyloxy-3-methoxybenzeneboronic acid, 98%, Thermo Scientific™

£47.60 - £165.00

Chemical Identifiers

CAS 243990-53-6
Molecular Formula C14H15BO4
Molecular Weight (g/mol) 258.08
MDL Number MFCD09027252
InChI Key PBVVDNNISDIUSU-UHFFFAOYSA-N
Synonym 4-benzyloxy-3-methoxyphenylboronic acid, 4-benzyloxy-3-methoxybenzeneboronic acid, 4-benzyloxy-3-methoxyphenyl boronic acid, 4-benzyloxoy-3-methoxyphenylboronic acid, acmc-209gbw, 4-benzyloxy-3-methoxyphenyl boronicacid, 4-benzyloxy-3-methoxybenzene boronic acid
PubChem CID 10682603
IUPAC Name (3-methoxy-4-phenylmethoxyphenyl)boronic acid
SMILES COC1=C(OCC2=CC=CC=C2)C=CC(=C1)B(O)O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15419207
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H52462.MD
250 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15409207
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H52462.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

243990-53-6
258.08
PBVVDNNISDIUSU-UHFFFAOYSA-N
10682603
COC1=C(OCC2=CC=CC=C2)C=CC(=C1)B(O)O
C14H15BO4
MFCD09027252
4-benzyloxy-3-methoxyphenylboronic acid, 4-benzyloxy-3-methoxybenzeneboronic acid, 4-benzyloxy-3-methoxyphenyl boronic acid, 4-benzyloxoy-3-methoxyphenylboronic acid, acmc-209gbw, 4-benzyloxy-3-methoxyphenyl boronicacid, 4-benzyloxy-3-methoxybenzene boronic acid
(3-methoxy-4-phenylmethoxyphenyl)boronic acid

Specifications

243990-53-6
C14H15BO4
4-benzyloxy-3-methoxyphenylboronic acid, 4-benzyloxy-3-methoxybenzeneboronic acid, 4-benzyloxy-3-methoxyphenyl boronic acid, 4-benzyloxoy-3-methoxyphenylboronic acid, acmc-209gbw, 4-benzyloxy-3-methoxyphenyl boronicacid, 4-benzyloxy-3-methoxybenzene boronic acid
COC1=C(OCC2=CC=CC=C2)C=CC(=C1)B(O)O
258.08
258.08
4-Benzyloxy-3-methoxybenzeneboronic acid
200°C to 203°C
MFCD09027252
PBVVDNNISDIUSU-UHFFFAOYSA-N
(3-methoxy-4-phenylmethoxyphenyl)boronic acid
10682603
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications